| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000587705.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000587705.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000587705.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000587705.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000587745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000587745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000592669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000592669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
ENST00000592669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006516.hg.1 |
ENST00000246914.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006516.hg.1 |
ENST00000316085.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006516.hg.1 |
ENST00000442804.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006516.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006516.hg.1 |
TC17000536.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700157725.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700157725.hg.1 |
ENST00000587705.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700157725.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700157725.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700157725.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700157725.hg.1 |
TC1700007915.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700157725.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700157725.hg.1 |
ENST00000587705.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700157725.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700157725.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700157725.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000246914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000587705.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000587705.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000587705.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000587705.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000587745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000587745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000591448.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000592072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000592669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000592669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
ENST00000592669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007915.hg.1 |
NM_032387.4 |
Refseq |