| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14008892.hg.1 |
ENST00000331952.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14008892.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14008892.hg.1 |
ENST00000553873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14008892.hg.1 |
ENST00000557649.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14008892.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14008892.hg.1 |
TC14000816.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1400160420.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400160420.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400160420.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400160420.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400160420.hg.1 |
TC1400008443.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008443.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400160423.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400160423.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400160423.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400160423.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400160423.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400160423.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400160423.hg.1 |
ENST00000331952.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400160423.hg.1 |
ENST00000410013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400160423.hg.1 |
ENST00000557649.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400160423.hg.1 |
NM_001008404.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400160423.hg.1 |
NM_001286399.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400160423.hg.1 |
NM_001286400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400160423.hg.1 |
TC1400008443.hg.1 |
Clariom D TC |