Assay Details
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011526675.2 | XP_011524977.1 | 6 - 7 | 628 | 104 | |
XM_011526676.2 | XP_011524978.1 | 6 - 7 | 593 | 104 | ||
XM_011526677.2 | XP_011524979.1 | 5 - 6 | 582 | 104 | ||
XM_011526678.2 | XP_011524980.1 | 6 - 7 | 574 | 104 | ||
XM_017026533.1 | XP_016882022.1 | 5 - 6 | 518 | 104 | ||
XM_017026534.1 | XP_016882023.1 | 5 - 6 | 491 | 104 | ||
GenBank mRNA | AK308939.1 | - | 1 - 2 | 132 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
signal transduction
positive regulation of cell proliferation
lymphocyte differentiation
regulation of myeloid dendritic cell activation
positive regulation of natural killer cell differentiation
embryonic hemopoiesis
positive regulation of myoblast differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
homeostasis of number of cells within a tissue
positive regulation of cell proliferation in bone marrow
negative regulation of apoptotic process in bone marrow
positive regulation of osteoclast proliferation
positive regulation of myoblast fusion
receptor binding
cytokine activity
receptor tyrosine kinase binding
protein homodimerization activity
positive regulation of protein phosphorylation
signal transduction
positive regulation of cell proliferation
lymphocyte differentiation
regulation of myeloid dendritic cell activation
positive regulation of natural killer cell differentiation
embryonic hemopoiesis
positive regulation of myoblast differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
homeostasis of number of cells within a tissue
positive regulation of cell proliferation in bone marrow
negative regulation of apoptotic process in bone marrow
positive regulation of osteoclast proliferation
positive regulation of myoblast fusion
receptor binding
cytokine activity
receptor tyrosine kinase binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000204637.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000204637.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000204637.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000204637.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000204637.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000204637.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000204637.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000204637.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000204637.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000204637.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000204637.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000204637.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000204637.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000593422.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000593422.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000593422.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000593422.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000593422.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000593422.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000593422.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000594009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000594009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000594009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000594009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000594009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000594009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000594009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000594009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000594009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000594009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000594009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000594009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000594009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000595510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000595510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000595510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000595510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000595510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000595510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000595510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000595510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000595510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000595510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000595510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000596435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000596435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000596435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000596435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000596435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000596435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000596435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000596435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000596435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000596435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000596435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000596435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000596435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000597914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000598472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000598472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000598472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000598472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000598472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000598472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000598555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000598555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000598555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000598555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000598555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000598555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000598555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600429.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600429.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600429.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600429.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600429.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600429.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600429.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600429.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600429.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600429.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600429.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600429.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000600429.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000601800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000601800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000601800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000601800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000601800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000601800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000601800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000601800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000601800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000601800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000601800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000601800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | ENST00000601800.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001204503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001278638.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011779.hg.1 | NM_001459.3 | Refseq |
