Assay Details
| Gene Symbol: TMEM263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319661.1 | NP_001306590.1 | 1 - 2 | 347 | 144 | |
| GenBank mRNA | AK022770.1 | - | 1 - 2 | 129 | 144 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK022770.1,NM_001319661.1 transcripts.
This assay is in the 5'UTR region of the AK022770.1,NM_001319661.1 transcripts.
Set Membership: |
|

TMEM263
Genomic Map