Assay Details
Gene Symbol: TMEM263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319661.1 | NP_001306590.1 | 2 - 3 | 414 | 101 | |
GenBank mRNA | AK022770.1 | - | 2 - 3 | 196 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK022770.1,NM_001319661.1 transcripts.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
