Assay Details
| Gene Symbol: STEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040665.1 | NP_001035755.1 | 4 - 5 | 1184 | 138 | |
| NM_001040666.1 | NP_001035756.1 | 4 - 5 | 1202 | 138 | ||
| NM_001244944.1 | NP_001231873.1 | 4 - 5 | 1521 | 138 | ||
| NM_001244945.1 | NP_001231874.1 | 3 - 4 | 1408 | 138 | ||
| NM_001244946.1 | NP_001231875.1 | 4 - 5 | 1184 | 138 | ||
| NM_152999.3 | NP_694544.2 | 3 - 4 | 1408 | 138 | ||
| XM_006715921.3 | XP_006715984.1 | 4 - 5 | 1518 | 138 | ||
| XM_017011953.1 | XP_016867442.1 | 4 - 5 | 1348 | 138 | ||
| XM_017011954.1 | XP_016867443.1 | 3 - 4 | 1235 | 138 | ||
| XM_017011955.1 | XP_016867444.1 | 4 - 5 | 1321 | 138 | ||
| XM_017011956.1 | XP_016867445.1 | 4 - 5 | 1519 | 138 | ||
| XM_017011957.1 | XP_016867446.1 | 4 - 5 | 1321 | 138 | ||
| XM_017011958.1 | XP_016867447.1 | 3 - 4 | 5183 | 138 | ||
| GenBank mRNA | AF455138.1 | - | 3 - 4 | 1408 | 138 | |
| AF526382.1 | - | 4 - 5 | 1202 | 138 | ||
| AK092666.1 | - | 3 - 4 | 1350 | 138 | ||
| AK315043.1 | - | 4 - 5 | 1322 | 138 | ||
| AY008443.1 | - | 4 - 5 | 1202 | 138 | ||
| AY008444.1 | - | 4 - 5 | 1184 | 138 | ||
| AY358267.1 | - | 4 - 5 | 1202 | 138 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

STEAP2
Genomic Map