| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041970.hg.1 |
ENST00000312233.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041970.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041970.hg.1 |
TC01002674.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041971.hg.1 |
ENST00000312233.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041971.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041971.hg.1 |
TC01002674.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100206616.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100206616.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100206616.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100206616.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100206616.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100206616.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100206616.hg.1 |
TC0100014240.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014240.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100206617.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100206617.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100206617.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100206617.hg.1 |
ENST00000312233.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100206617.hg.1 |
ENST00000628545.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100206617.hg.1 |
NM_147193.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100206617.hg.1 |
TC0100014240.hg.1 |
Clariom D TC |