Assay Details
| Gene Symbol: FAM24A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029888.1 | NP_001025059.1 | 2 - 3 | 239 | 77 | |
| XM_017015638.1 | XP_016871127.1 | 2 - 3 | 342 | 77 | ||
| XM_017015639.1 | XP_016871128.1 | 2 - 3 | 247 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000894.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929014 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538188 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538240 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538204 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538187 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929052 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929043 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929015 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538181 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538180 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929013 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538185 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538184 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538209 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929035 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16392195 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929033 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538182 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929041 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16392192 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538194 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929011 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538201 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929036 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538215 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538234 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538241 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538192 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538233 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538228 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538189 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538208 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538214 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538216 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929047 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929040 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929005 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538211 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538222 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538230 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538213 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07538218 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16392191 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27929054 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P865708 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P865708 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 6428 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 8214 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P32791017 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P02361877 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P865708 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P865708 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929033 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538216 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929015 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929013 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929009 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929005 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538188 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929023 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538184 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929012 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929014 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929036 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538199 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929005 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929012 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929033 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538188 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929015 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929014 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538184 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929036 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538199 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929013 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929009 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929023 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538216 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27929040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07538209 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P865708 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P865708 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19096206 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P865708 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538205 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538182 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538202 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538184 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16392195 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16392190 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16392193 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538195 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538215 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538234 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16392189 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538226 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538229 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538199 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538230 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538216 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538224 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538220 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538191 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538194 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538218 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538238 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538228 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538193 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538197 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16392192 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538186 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538232 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538208 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538235 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538180 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538188 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538213 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07538211 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001029888_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7931156 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7931157 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7931158 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7931155 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 243143_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11733719_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11733719_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009164.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009164.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009164.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009164.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009164.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009164.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009164.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009164.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10010949.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10010949.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10010949.hg.1 | TC10000894.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10010951.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10010951.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10010951.hg.1 | TC10000894.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10010950.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10010950.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10010950.hg.1 | TC10000894.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000009164.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009164.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009164.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009164.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009164.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009164.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009164.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009164.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162895.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162895.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162895.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162895.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162895.hg.1 | TC1000009164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000162894.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162894.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162894.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162894.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162894.hg.1 | TC1000009164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000162896.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162896.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162896.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162896.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162896.hg.1 | TC1000009164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000162892.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162892.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162892.hg.1 | ENST00000368894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162892.hg.1 | NM_001029888.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162892.hg.1 | TC1000009164.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 243143_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 243143_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_001029888_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243143_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243143_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243143_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243143_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243143_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243143_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243143_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243143_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243143_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243143_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243143_at4 | - | - |
| Human Genome U95 Set | 42510_PM_at | - | - |
| Human Genome U95 Set | 42510_at | - | - |
| Human Genome U133 Set | 243143_at | - | - |
| Human Genome U133 Set | 243143_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044603 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044603 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 130928 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1783969 | NM_001029888.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1783969 | NM_001029888.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1783969 | NM_001029888.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1783969 | NM_001029888.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1783969 | NM_001029888.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_4837-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1783969 | NM_001029888.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1783969 | NM_001029888.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 4280377 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_4837 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1783969 | NM_001029888.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1783969 | NM_001029888.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1783969 | NM_001029888.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1783969 | NM_001029888.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1783969 | NM_001029888.1 | human_transcriptome |

FAM24A
Genomic Map