Assay Details
| Gene Symbol: ARSF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201538.1 | NP_001188467.1 | 7 - 8 | 1105 | 95 | |
| NM_001201539.1 | NP_001188468.1 | 7 - 8 | 1190 | 95 | ||
| NM_004042.4 | NP_004033.2 | 7 - 8 | 1071 | 95 | ||
| XM_011545522.1 | XP_011543824.1 | 7 - 8 | 1057 | 95 | ||
| GenBank mRNA | BC022389.1 | - | 7 - 8 | 1190 | 4609825 | 95 |
| X97868.1 | - | 8 - 9 | 1042 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000020.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000319.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000319.hg.1 | ENST00000381127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000319.hg.1 | ENST00000537104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000319.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000319.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000319.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000319.hg.1 | TC0X000020.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000318.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000318.hg.1 | ENST00000381127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000318.hg.1 | ENST00000537104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000318.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000318.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000318.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000318.hg.1 | TC0X000020.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144555.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144555.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144555.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144555.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144555.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144555.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144555.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144555.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144555.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144555.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144555.hg.1 | TC0X00006489.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00144554.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144554.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144554.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144554.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144554.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144554.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144554.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144554.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144554.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144554.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144554.hg.1 | TC0X00006489.hg.1 | Clariom D TC |

ARSF
Genomic Map