Assay Details
| Gene Symbol: ARSF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201538.1 | NP_001188467.1 | 8 - 9 | 1242 | 74 | |
| NM_001201539.1 | NP_001188468.1 | 8 - 9 | 1327 | 74 | ||
| NM_004042.4 | NP_004033.2 | 8 - 9 | 1208 | 74 | ||
| XM_011545522.1 | XP_011543824.1 | 8 - 9 | 1194 | 74 | ||
| XM_011545523.1 | XP_011543825.1 | 8 - 9 | 1107 | 74 | ||
| XM_017029527.1 | XP_016885016.1 | 8 - 9 | 1121 | 74 | ||
| GenBank mRNA | BC022389.1 | - | 8 - 9 | 1327 | 4609825 | 74 |
| X97868.1 | - | 9 - 10 | 1179 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000020.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000320.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000320.hg.1 | ENST00000381127.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000320.hg.1 | ENST00000537104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000320.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000320.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000320.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000320.hg.1 | TC0X000020.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144556.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144556.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144556.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144556.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144556.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144556.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144556.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144556.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144556.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144556.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144556.hg.1 | TC0X00006489.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000359361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | ENST00000381127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201538.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_001201539.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006489.hg.1 | NM_004042.4 | Refseq |

ARSF
Genomic Map