Assay Details
| Gene Symbol: APBB1IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019043.3 | NP_061916.3 | 12 - 13 | 1581 | 64 | |
| XM_006717451.2 | XP_006717514.1 | 11 - 12 | 1279 | 64 | ||
| XM_011519514.2 | XP_011517816.1 | 11 - 12 | 1450 | 64 | ||
| GenBank mRNA | AK303597.1 | - | 12 - 13 | 2141 | 64 | |
| AY152730.1 | - | 11 - 12 | 1347 | 64 | ||
| BC054516.1 | - | 11 - 12 | 1359 | 5172946 | 64 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000181.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10002168.hg.1 | ENST00000376236.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002168.hg.1 | ENST00000445780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002168.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002168.hg.1 | TC10000181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002172.hg.1 | ENST00000376236.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002172.hg.1 | ENST00000445780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002172.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002172.hg.1 | TC10000181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000362863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000493857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000493857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | ENST00000493857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002171.hg.1 | ENST00000376236.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002171.hg.1 | ENST00000445780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002171.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002171.hg.1 | TC10000181.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000148253.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148253.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148253.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148253.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148253.hg.1 | TC1000007072.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148251.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148251.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148251.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148251.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148251.hg.1 | TC1000007072.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148252.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148252.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148252.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148252.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148252.hg.1 | TC1000007072.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000356785.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000362863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000376236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000493857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000493857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | ENST00000493857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007072.hg.1 | NM_019043.3 | Refseq |

APBB1IP
Genomic Map