| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021875.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021875.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021875.hg.1 |
TC06001810.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021873.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021873.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021873.hg.1 |
TC06001810.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021874.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021874.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021874.hg.1 |
TC06001810.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600183075.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183075.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183075.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183075.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183075.hg.1 |
TC0600012064.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012064.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183076.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183076.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183076.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183076.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183076.hg.1 |
TC0600012064.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600183077.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183077.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183077.hg.1 |
ENST00000259803.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183077.hg.1 |
NM_003643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183077.hg.1 |
TC0600012064.hg.1 |
Clariom D TC |