| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000426125.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000426125.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000426125.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000427819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000427819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000427819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000427819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000427819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000427819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000427819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000427819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000427819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000427819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000427819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
ENST00000427819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
NR_045484.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
NR_045484.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
NR_045484.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
NR_045484.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008907.hg.1 |
NR_045484.1 |
Refseq |