| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07016426.hg.1 |
ENST00000354513.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07016426.hg.1 |
ENST00000402802.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07016426.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07016426.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07016426.hg.1 |
TC07001073.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000426681.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000426681.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000482462.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000482462.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000482462.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000482462.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
ENST00000482462.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000426681.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000426681.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000482462.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000482462.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000482462.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000482462.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
ENST00000482462.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009977.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
ENST00000482462.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
ENST00000354513.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
ENST00000400761.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
ENST00000402802.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
ENST00000405692.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
ENST00000482462.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
NM_002607.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
NM_033023.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700170974.hg.1 |
TC0700009977.hg.1 |
Clariom D TC |