| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
ENST00000393952.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
ENST00000393952.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
ENST00000393952.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
ENST00000393952.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
ENST00000531590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
ENST00000531590.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
ENST00000309602.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
ENST00000393952.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
ENST00000393952.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
ENST00000393952.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
ENST00000393952.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
ENST00000531590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
ENST00000531590.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008102.hg.1 |
NM_024036.4 |
Refseq |