| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000550203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000550203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000550203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000550203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12023824.hg.1 |
ENST00000261172.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12023824.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12023824.hg.1 |
TC12001801.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000550203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000550203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000550203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000550203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011491.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189036.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189036.hg.1 |
ENST00000550203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189036.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189036.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189036.hg.1 |
ENST00000261172.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189036.hg.1 |
ENST00000550203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189036.hg.1 |
ENST00000551767.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189036.hg.1 |
NM_004950.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189036.hg.1 |
TC1200011491.hg.1 |
Clariom D TC |