Assay Details
| Gene Symbol: NDUFA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002488.4 | NP_002479.1 | 2 - 3 | 412 | 77 | |
| NR_033697.1 | 1 - 2 | 532 | 77 | |||
| GenBank mRNA | AF047185.1 | - | 2 - 3 | 266 | 77 | |
| AF077029.1 | - | 2 - 3 | 283 | 77 | ||
| BC003674.1 | - | 2 - 3 | 257 | 4044385 | 77 | |
| CR457016.1 | - | 2 - 3 | 210 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001862.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05025586.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025586.hg.1 | TC05001862.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000510680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000510680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000510680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000510680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012248.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185109.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185109.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185109.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185109.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185109.hg.1 | TC0500012248.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | ENST00000510680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | ENST00000252102.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | ENST00000502960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | ENST00000510680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | ENST00000512088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | NM_001185012.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | NM_002488.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | NR_033697.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500185107.hg.1 | TC0500012248.hg.1 | Clariom D TC |

NDUFA2
Genomic Map