Assay Details
| Gene Symbol: EBF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022659.3 | NP_073150.2 | 9 - 10 | 1144 | 91 | |
| GenBank mRNA | AK021562.1 | - | 2 - 3 | 183 | 91 | |
| AK304372.1 | - | 8 - 9 | 792 | 91 | ||
| BC069665.1 | - | 2 - 3 | 161 | 91 | ||
| BC069726.1 | - | 2 - 3 | 164 | 91 | ||
| BC069747.1 | - | 2 - 3 | 163 | 91 | ||
| BC069768.1 | - | 2 - 3 | 164 | 91 | ||
| BC074794.2 | - | 1 - 2 | 100 | 91 | ||
| BC113478.1 | - | 1 - 2 | 128 | 91 | ||
| BC113504.1 | - | 1 - 2 | 128 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001073.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell fate determination
transcription from RNA polymerase II promoter
positive regulation of chromatin binding
positive regulation of transcription from RNA polymerase II promoter
brown fat cell differentiation
adipose tissue development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
metal ion binding
cell fate determination
transcription from RNA polymerase II promoter
positive regulation of chromatin binding
positive regulation of transcription from RNA polymerase II promoter
brown fat cell differentiation
adipose tissue development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
metal ion binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08013643.hg.1 | ENST00000408929.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013643.hg.1 | ENST00000520164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013643.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013643.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013643.hg.1 | TC08001073.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013642.hg.1 | ENST00000408929.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013642.hg.1 | ENST00000520164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013642.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013642.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013642.hg.1 | TC08001073.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013644.hg.1 | ENST00000408929.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013644.hg.1 | ENST00000520164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013644.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013644.hg.1 | TC08001073.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167397.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167397.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167397.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167397.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167397.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167397.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167397.hg.1 | TC0800009927.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000517825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009927.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167395.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167395.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167395.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167395.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167395.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167395.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167395.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167395.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167395.hg.1 | TC0800009927.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800167396.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167396.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167396.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167396.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167396.hg.1 | ENST00000408929.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167396.hg.1 | ENST00000520164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167396.hg.1 | ENST00000535548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167396.hg.1 | NM_022659.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167396.hg.1 | TC0800009927.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 114799 | - | - |

EBF2
Genomic Map