| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009627.hg.1 |
ENST00000245323.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009627.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009627.hg.1 |
TC13000860.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009629.hg.1 |
ENST00000245323.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009629.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009629.hg.1 |
TC13000860.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009626.hg.1 |
ENST00000245323.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009626.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009626.hg.1 |
TC13000860.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164886.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164886.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164886.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164886.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164886.hg.1 |
TC1300009712.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300164890.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164890.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164890.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164890.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164890.hg.1 |
TC1300009712.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300164887.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164887.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164887.hg.1 |
ENST00000245323.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300164887.hg.1 |
NM_004093.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300164887.hg.1 |
TC1300009712.hg.1 |
Clariom D TC |