Assay Details
| Gene Symbol: RHPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052924.2 | NP_443156.2 | 11 - 12 | 1548 | 77 | |
| XM_005250772.3 | XP_005250829.1 | 12 - 13 | 1734 | 77 | ||
| XM_005250773.3 | XP_005250830.1 | 12 - 13 | 1731 | 77 | ||
| XM_006716502.3 | XP_006716565.1 | 12 - 13 | 1749 | 77 | ||
| XM_011516817.2 | XP_011515119.1 | 12 - 13 | 1752 | 77 | ||
| XM_011516818.1 | XP_011515120.1 | 12 - 13 | 2409 | 77 | ||
| XM_011516819.2 | XP_011515121.1 | 11 - 12 | 1593 | 77 | ||
| XM_011516820.2 | XP_011515122.1 | 11 - 12 | 1936 | 77 | ||
| XM_011516821.2 | XP_011515123.1 | 11 - 12 | 1940 | 77 | ||
| XM_011516822.2 | XP_011515124.1 | 11 - 12 | 2085 | 77 | ||
| XM_017013013.1 | XP_016868502.1 | 11 - 12 | 1624 | 77 | ||
| XM_017013015.1 | XP_016868504.1 | 11 - 12 | 2066 | 77 | ||
| GenBank mRNA | AB067516.1 | - | 10 - 11 | 1482 | 77 | |
| AY082588.1 | - | 10 - 11 | 1490 | 77 | ||
| BC025767.1 | - | 11 - 12 | 1516 | 5211354 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000820.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08010897.hg.1 | ENST00000289013.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010897.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010897.hg.1 | TC08000820.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010898.hg.1 | ENST00000289013.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010898.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010898.hg.1 | TC08000820.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0800162591.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162591.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162591.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162591.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162591.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800162591.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162591.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162591.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162591.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162591.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800162591.hg.1 | TC0800009194.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800162590.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162590.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162590.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162590.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162590.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800162590.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162590.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162590.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162590.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162590.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800162590.hg.1 | TC0800009194.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |

RHPN1
Genomic Map