Assay Details
| Gene Symbol: RHPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052924.2 | NP_443156.2 | 1 - 2 | 195 | 147 | |
| XM_011516819.2 | XP_011515121.1 | 1 - 2 | 222 | 147 | ||
| GenBank mRNA | BC025767.1 | - | 1 - 2 | 163 | 5211354 | 147 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000820.hg
Additional Information:
This assay is in the 5'UTR region of the BC025767.1,NM_052924.2,XM_011516819.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08010882.hg.1 | ENST00000289013.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010882.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010882.hg.1 | TC08000820.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010881.hg.1 | ENST00000289013.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010881.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010881.hg.1 | TC08000820.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010883.hg.1 | ENST00000289013.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010883.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010883.hg.1 | TC08000820.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010880.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010880.hg.1 | TC08000820.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0800162576.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162576.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162576.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162576.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800162576.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162576.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162576.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162576.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800162576.hg.1 | TC0800009194.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800162575.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162575.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162575.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162575.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800162575.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162575.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162575.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162575.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800162575.hg.1 | TC0800009194.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800162573.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162573.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800162573.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162573.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800162573.hg.1 | TC0800009194.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000522899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009194.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800162574.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162574.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162574.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800162574.hg.1 | ENST00000289013.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162574.hg.1 | ENST00000620174.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800162574.hg.1 | NM_052924.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800162574.hg.1 | TC0800009194.hg.1 | Clariom D TC |

RHPN1
Genomic Map