Assay Details
| Gene Symbol: ZNF444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005259036.1 | XP_005259093.1 | 1 - 2 | 95 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005259036.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000587664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000337080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000586123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587195.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587467.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000587664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000588358.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000591564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000592949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | ENST00000593100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_001253792.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008943.hg.1 | NM_018337.3 | Refseq |

ZNF444
Genomic Map