Assay Details
| Gene Symbol: SLX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032444.2 | NP_115820.2 | 7 - 8 | 2310 | 126 | |
| XM_011522715.2 | XP_011521017.1 | 7 - 8 | 2324 | 126 | ||
| XM_017023775.1 | XP_016879264.1 | 4 - 5 | 1715 | 126 | ||
| GenBank mRNA | AB075867.1 | - | 5 - 6 | 2904 | 126 | |
| BC029952.1 | - | 6 - 7 | 1754 | 126 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000821.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
DNA replication
DNA repair
nucleotide-excision repair
DNA double-strand break processing involved in repair via single-strand annealing
interstrand cross-link repair
positive regulation of catalytic activity
response to intra-S DNA damage checkpoint signaling
t-circle formation
positive regulation of t-circle formation
endodeoxyribonuclease activity
protein binding
enzyme activator activity
crossover junction endodeoxyribonuclease activity
5'-flap endonuclease activity
3'-flap endonuclease activity
double-strand break repair via homologous recombination
DNA replication
DNA repair
nucleotide-excision repair
DNA double-strand break processing involved in repair via single-strand annealing
interstrand cross-link repair
positive regulation of catalytic activity
response to intra-S DNA damage checkpoint signaling
t-circle formation
positive regulation of t-circle formation
endodeoxyribonuclease activity
protein binding
enzyme activator activity
crossover junction endodeoxyribonuclease activity
5'-flap endonuclease activity
3'-flap endonuclease activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16011871.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011871.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011871.hg.1 | TC16000821.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170231.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170231.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170231.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170231.hg.1 | TC1600009199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600170231.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170231.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170231.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |

SLX4
Genomic Map