Assay Details
| Gene Symbol: SLX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032444.2 | NP_115820.2 | 3 - 4 | 1392 | 85 | |
| XM_011522715.2 | XP_011521017.1 | 3 - 4 | 1406 | 85 | ||
| GenBank mRNA | AB075867.1 | - | 2 - 3 | 1060 | 85 | |
| AK122775.1 | - | 3 - 4 | 1393 | 85 | ||
| BC029952.1 | - | 2 - 3 | 836 | 85 | ||
| BC104222.2 | - | 2 - 3 | 85 | |||
| BC104223.2 | - | 2 - 3 | 271 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000821.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB075867.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
DNA replication
DNA repair
nucleotide-excision repair
DNA double-strand break processing involved in repair via single-strand annealing
interstrand cross-link repair
positive regulation of catalytic activity
response to intra-S DNA damage checkpoint signaling
t-circle formation
positive regulation of t-circle formation
endodeoxyribonuclease activity
protein binding
enzyme activator activity
crossover junction endodeoxyribonuclease activity
5'-flap endonuclease activity
3'-flap endonuclease activity
double-strand break repair via homologous recombination
DNA replication
DNA repair
nucleotide-excision repair
DNA double-strand break processing involved in repair via single-strand annealing
interstrand cross-link repair
positive regulation of catalytic activity
response to intra-S DNA damage checkpoint signaling
t-circle formation
positive regulation of t-circle formation
endodeoxyribonuclease activity
protein binding
enzyme activator activity
crossover junction endodeoxyribonuclease activity
5'-flap endonuclease activity
3'-flap endonuclease activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011877.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011877.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011877.hg.1 | TC16000821.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011880.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011880.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011880.hg.1 | TC16000821.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011878.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011878.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011878.hg.1 | TC16000821.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011879.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011879.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011879.hg.1 | TC16000821.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600170240.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170240.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170240.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170240.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170240.hg.1 | TC1600009199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600170240.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170240.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170240.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170240.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170237.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170237.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170237.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170237.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170237.hg.1 | TC1600009199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600170237.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170237.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170237.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170237.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009199.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170239.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170239.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170239.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170239.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170239.hg.1 | TC1600009199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600170239.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170239.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170239.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170239.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170241.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170241.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170241.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170241.hg.1 | TC1600009199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600170241.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170241.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170241.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170242.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170242.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170242.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170242.hg.1 | TC1600009199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600170242.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170242.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170242.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170238.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170238.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170238.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170238.hg.1 | NM_032444.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170238.hg.1 | TC1600009199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600170238.hg.1 | ENST00000294008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170238.hg.1 | ENST00000466154.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170238.hg.1 | ENST00000486524.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170238.hg.1 | NM_032444.3 | Refseq |

SLX4
Genomic Map