| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000587351.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000587351.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000587351.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004098.hg.1 |
ENST00000263384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004098.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004098.hg.1 |
TC19000319.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004099.hg.1 |
ENST00000263384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004099.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004099.hg.1 |
TC19000319.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006511.hg.1 |
ENST00000344033.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006511.hg.1 |
ENST00000344033.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006511.hg.1 |
ENST00000344033.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010683.hg.1 |
ENST00000603244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152650.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152650.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152650.hg.1 |
ENST00000587351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152650.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152650.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152650.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900152650.hg.1 |
TC1900007265.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900152650.hg.1 |
ENST00000587351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152650.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152650.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152650.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152650.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152650.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000587351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000587351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000587351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007265.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900152652.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152652.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152652.hg.1 |
ENST00000587351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152652.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152652.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152652.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900152652.hg.1 |
TC1900007265.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900152652.hg.1 |
ENST00000263384.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152652.hg.1 |
ENST00000585831.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152652.hg.1 |
ENST00000587351.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152652.hg.1 |
ENST00000588367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152652.hg.1 |
ENST00000589852.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900152652.hg.1 |
NM_014077.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010683.hg.1 |
ENST00000603244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006511.hg.1 |
ENST00000344033.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006511.hg.1 |
ENST00000344033.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006511.hg.1 |
ENST00000344033.5 |
Ensembl |