| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000570065.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
ENST00000570065.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16007018.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16007018.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16007018.hg.1 |
TC16000500.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1600159214.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159214.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600159214.hg.1 |
TC1600008021.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600159214.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600159214.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000219271.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000570065.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
ENST00000570065.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008021.hg.1 |
NM_002428.3 |
Refseq |