Assay Details
| Gene Symbol: MOK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017021551.1 | XP_016877040.1 | 2 - 3 | 256 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017021551.1 transcripts.
Set Membership: |
|

MOK
Genomic Map