Assay Details
| Gene Symbol: SYNJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178088.1 | NP_001171559.1 | 1 - 2 | 236 | 98 | |
| NM_003898.3 | NP_003889.1 | 2 - 3 | 313 | 98 | ||
| XM_006715592.3 | XP_006715655.1 | 2 - 3 | 313 | 98 | ||
| XM_011536224.1 | XP_011534526.1 | 2 - 3 | 313 | 98 | ||
| XM_011536225.1 | XP_011534527.1 | 2 - 3 | 353 | 98 | ||
| XM_011536227.2 | XP_011534529.1 | 2 - 3 | 313 | 98 | ||
| XM_011536230.2 | XP_011534532.1 | 2 - 3 | 313 | 98 | ||
| GenBank mRNA | AF318616.1 | - | 2 - 3 | 290 | 98 | |
| AK294480.1 | - | 2 - 3 | 313 | 98 | ||
| AY152396.1 | - | 2 - 3 | 215 | 98 | ||
| BX648749.1 | - | 1 - 2 | 143 | 98 | ||
| DA327619.1 | - | 1 - 2 | 236 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001128.hg
Additional Information:
This assay is in the 5'UTR region of the NM_001178088.1,XM_011536225.1 transcripts.
Set Membership: |
|

SYNJ2
Genomic Map