| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
ENST00000296266.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
ENST00000347300.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
ENST00000348417.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
ENST00000349441.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
ENST00000431818.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
ENST00000440957.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
ENST00000446384.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
ENST00000454840.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
ENST00000504021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
ENST00000507564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
NM_018262.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
NM_052985.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
NM_052989.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
NM_052990.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012404.hg.1 |
TC03000692.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000296266.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000347300.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000348417.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000349441.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000431818.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000440957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000504021.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000506507.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000507564.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000511425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000512220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_001280541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_001280545.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_001280546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_018262.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_052985.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_052989.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_052990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000296266.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000347300.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000348417.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000349441.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000431818.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000440957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000504021.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000506507.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000507564.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000511425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
ENST00000512220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_001280541.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_001280545.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_001280546.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_018262.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_052985.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_052989.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
NM_052990.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162485.hg.1 |
TC0300008782.hg.1 |
Clariom D TC |