Assay Details
| Gene Symbol: ZNF662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134656.1 | NP_001128128.1 | 1 - 2 | 278 | 88 | |
| GenBank mRNA | AK128676.1 | - | 1 - 2 | 270 | 88 | |
| AW117418.1 | - | 1 - 2 | 290 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03003345.hg
Additional Information:
This assay is in the 5'UTR region of the AK128676.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03004576.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004576.hg.1 | ENST00000541208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004576.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004576.hg.1 | TC03003345.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000430067.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000430067.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000430067.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004578.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004578.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004578.hg.1 | ENST00000440367.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004578.hg.1 | ENST00000541208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004578.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004578.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004578.hg.1 | TC03003345.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000430067.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000430067.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000430067.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203848.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203848.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203848.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203848.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203848.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203848.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203848.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203848.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203848.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203848.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203848.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203848.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203848.hg.1 | TC0300013814.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300203845.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203845.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203845.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203845.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203845.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203845.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203845.hg.1 | TC0300013814.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300203846.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203846.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203846.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203846.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203846.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203846.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203846.hg.1 | TC0300013814.hg.1 | Clariom D TC |

ZNF662
Genomic Map