Assay Details
| Gene Symbol: ZNF662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134656.1 | NP_001128128.1 | 3 - 4 | 515 | 128 | |
| GenBank mRNA | AK128676.1 | - | 2 - 3 | 1154 | 128 | |
| BC128085.1 | - | 2 - 3 | 228 | 40108370 | 128 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03003345.hg
Additional Information:
This assay is in the 5'UTR region of the AK128676.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03004587.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004587.hg.1 | ENST00000440367.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004587.hg.1 | ENST00000541208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004587.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004587.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004587.hg.1 | TC03003345.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004591.hg.1 | ENST00000440367.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004591.hg.1 | ENST00000541208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004591.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004591.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004591.hg.1 | TC03003345.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000430067.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000430067.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000430067.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004590.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004590.hg.1 | ENST00000440367.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004590.hg.1 | ENST00000541208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004590.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004590.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004590.hg.1 | TC03003345.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004594.hg.1 | ENST00000541208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004594.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004594.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004594.hg.1 | TC03003345.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004588.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004588.hg.1 | ENST00000440367.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004588.hg.1 | ENST00000541208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004588.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004588.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004588.hg.1 | TC03003345.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004592.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004592.hg.1 | ENST00000440367.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004592.hg.1 | ENST00000541208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004592.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004592.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004592.hg.1 | TC03003345.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300203860.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203860.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203860.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203860.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203860.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203860.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203860.hg.1 | TC0300013814.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000430067.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000430067.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000430067.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013814.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203859.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203859.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203859.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203859.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203859.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203859.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203859.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203859.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203859.hg.1 | TC0300013814.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300203861.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203861.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203861.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203861.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203861.hg.1 | ENST00000422021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203861.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203861.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203861.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203861.hg.1 | TC0300013814.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300203863.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203863.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203863.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203863.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203863.hg.1 | TC0300013814.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300203857.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203857.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203857.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203857.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203857.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203857.hg.1 | ENST00000328199.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203857.hg.1 | ENST00000440367.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203857.hg.1 | ENST00000475386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300203857.hg.1 | NM_001134656.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203857.hg.1 | NM_207404.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300203857.hg.1 | TC0300013814.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 102916 | - | - |

ZNF662
Genomic Map