Assay Details
| Gene Symbol: ZNF652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145365.1 | NP_001138837.1 | 3 - 4 | 1516 | 67 | |
| NM_014897.2 | NP_055712.1 | 3 - 4 | 1371 | 67 | ||
| NR_135579.1 | 4 - 5 | 1100 | 67 | |||
| XM_005257166.2 | XP_005257223.1 | 2 - 3 | 723 | 67 | ||
| GenBank mRNA | AB023141.1 | - | 3 - 4 | 1371 | 67 | |
| AK292667.1 | - | 3 - 4 | 1506 | 67 | ||
| BC139779.1 | - | 2 - 3 | 1226 | 40122232 | 67 | |
| BC152440.1 | - | 3 - 4 | 1371 | 8862563 | 67 | |
| CR933680.1 | - | 3 - 4 | 1367 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001662.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17022272.hg.1 | ENST00000362063.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022272.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022272.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022272.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022272.hg.1 | TC17001662.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000513488.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000513488.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000513488.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000513488.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188811.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188811.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188811.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188811.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188811.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188811.hg.1 | TC1700011038.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700188811.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188811.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188811.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188811.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188811.hg.1 | NM_014897.2 | Refseq |

ZNF652
Genomic Map