Assay Details
| Gene Symbol: CHMP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024591.4 | NP_078867.2 | 2 - 3 | 256 | 125 | |
| XM_005257668.1 | XP_005257725.1 | 2 - 3 | 256 | 125 | ||
| GenBank mRNA | AK021811.1 | - | 2 - 3 | 194 | 125 | |
| AK223187.1 | - | 2 - 3 | 218 | 125 | ||
| AK292105.1 | - | 2 - 3 | 222 | 125 | ||
| AY329087.1 | - | 2 - 3 | 178 | 125 | ||
| BC010108.2 | - | 2 - 3 | 202 | 3604039 | 125 | |
| BP196406.1 | - | 2 - 3 | 256 | 125 | ||
| CR457284.1 | - | 2 - 3 | 178 | 125 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000922.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell separation after cytokinesis
autophagy
nucleus organization
mitotic metaphase plate congression
protein transport
endosomal transport
viral life cycle
multivesicular body assembly
viral budding via host ESCRT complex
regulation of exosomal secretion
ESCRT III complex assembly
protein binding
protein N-terminus binding
cell separation after cytokinesis
autophagy
nucleus organization
mitotic metaphase plate congression
protein transport
endosomal transport
viral life cycle
multivesicular body assembly
viral budding via host ESCRT complex
regulation of exosomal secretion
ESCRT III complex assembly
protein binding
protein N-terminus binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012131.hg.1 | ENST00000325167.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012131.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012131.hg.1 | TC17000922.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012130.hg.1 | ENST00000325167.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012130.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012130.hg.1 | TC17000922.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012132.hg.1 | ENST00000325167.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012132.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012132.hg.1 | TC17000922.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012133.hg.1 | ENST00000325167.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012133.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012133.hg.1 | TC17000922.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009144.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169289.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169289.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169289.hg.1 | TC1700009144.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700169289.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169289.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169291.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169291.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169291.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169291.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169291.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169291.hg.1 | TC1700009144.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700169291.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169291.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169291.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169291.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169291.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169290.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169290.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169290.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169290.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169290.hg.1 | TC1700009144.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700169290.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169290.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169290.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169290.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169292.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169292.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169292.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169292.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169292.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169292.hg.1 | TC1700009144.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700169292.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169292.hg.1 | ENST00000571457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169292.hg.1 | ENST00000572525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169292.hg.1 | ENST00000572778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169292.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169287.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169287.hg.1 | NM_024591.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169287.hg.1 | TC1700009144.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700169287.hg.1 | ENST00000325167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169287.hg.1 | NM_024591.4 | Refseq |

CHMP6
Genomic Map