| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012136.hg.1 |
ENST00000325167.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012136.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012136.hg.1 |
TC17000922.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012137.hg.1 |
ENST00000325167.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012137.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012137.hg.1 |
TC17000922.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700169295.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169295.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169295.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169295.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169295.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169295.hg.1 |
TC1700009144.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700169295.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169295.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169295.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169295.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169295.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009144.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169296.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169296.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169296.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169296.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169296.hg.1 |
NM_024591.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169296.hg.1 |
TC1700009144.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700169296.hg.1 |
ENST00000325167.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169296.hg.1 |
ENST00000571457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169296.hg.1 |
ENST00000572525.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169296.hg.1 |
ENST00000572778.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169296.hg.1 |
NM_024591.4 |
Refseq |