Assay Details
| Gene Symbol: MCUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031713.3 | NP_001026883.1 | 3 - 4 | 767 | 132 | |
| XM_011514802.1 | XP_011513104.1 | 3 - 4 | 770 | 132 | ||
| GenBank mRNA | AK024611.1 | - | 4 - 5 | 1362 | 132 | |
| BC008234.1 | - | 3 - 4 | 522 | 132 | ||
| BC016850.1 | - | 3 - 4 | 741 | 3886144 | 132 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001282.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009424.hg.1 | ENST00000573670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015409.hg.1 | ENST00000379170.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015409.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015409.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015409.hg.1 | TC06001282.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0600173869.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173869.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173869.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173869.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173869.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173869.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173869.hg.1 | TC0600010867.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000379170.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000488770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | ENST00000607303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010867.hg.1 | NM_001031713.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009424.hg.1 | ENST00000573670.1 | Ensembl |

MCUR1
Genomic Map