| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015713.hg.1 |
ENST00000355221.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015713.hg.1 |
ENST00000359530.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015713.hg.1 |
ENST00000396774.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015713.hg.1 |
ENST00000520188.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015713.hg.1 |
NM_015941.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015713.hg.1 |
NM_213619.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015713.hg.1 |
NM_213620.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015713.hg.1 |
TC08001215.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015714.hg.1 |
ENST00000355221.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015714.hg.1 |
ENST00000359530.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015714.hg.1 |
ENST00000396774.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015714.hg.1 |
ENST00000520188.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015714.hg.1 |
NM_015941.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015714.hg.1 |
NM_213619.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015714.hg.1 |
NM_213620.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08015714.hg.1 |
TC08001215.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000355221.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000359530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000396774.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000519588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000520188.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000521377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000521900.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000522849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000524164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
NM_015941.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
NM_213619.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
NM_213620.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000355221.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000359530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000396774.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000519588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000520188.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000521377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000521900.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000522849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
ENST00000524164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
NM_015941.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
NM_213619.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
NM_213620.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170998.hg.1 |
TC0800010450.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000355221.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000359530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000396774.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000520188.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000521377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000521900.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000522849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000524164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
NM_015941.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
NM_213619.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
NM_213620.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000355221.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000359530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000396774.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000520188.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000521377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000521900.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000522849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
ENST00000524164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
NM_015941.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
NM_213619.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
NM_213620.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170997.hg.1 |
TC0800010450.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000355221.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000359530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000396774.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000519588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000520188.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000521377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000521900.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000522849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000524164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
NM_015941.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
NM_213619.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
NM_213620.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000355221.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000359530.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000396774.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000519588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000520188.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000521377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000521900.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000522849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
ENST00000524164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
NM_015941.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
NM_213619.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
NM_213620.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800170999.hg.1 |
TC0800010450.hg.1 |
Clariom D TC |