Assay Details
| Gene Symbol: P2RY10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324218.1 | NP_001311147.1 | 3 | 1834 | 92 | |
| NM_001324221.1 | NP_001311150.1 | 3 | 1740 | 92 | ||
| NM_001324225.1 | NP_001311154.1 | 4 | 1819 | 92 | ||
| NM_014499.3 | NP_055314.1 | 4 | 1883 | 92 | ||
| NM_198333.2 | NP_938147.1 | 2 | 1691 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P2RY10
Genomic Map