| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03013425.hg.1 |
ENST00000183605.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03013425.hg.1 |
ENST00000343735.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03013425.hg.1 |
ENST00000536138.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03013425.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03013425.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03013425.hg.1 |
TC03000742.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000479660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000479660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000479660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000479660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
ENST00000479660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300163918.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300163918.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300163918.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300163918.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300163918.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300163918.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300163918.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300163918.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300163918.hg.1 |
TC0300008933.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000183605.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000343735.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000479660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000479660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000479660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000479660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
ENST00000479660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_001002026.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008933.hg.1 |
NM_016369.3 |
Refseq |