Assay Details
| Gene Symbol: MSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018133.3 | NP_060603.2 | 1 - 2 | 872 | 64 | |
| GenBank mRNA | AB046805.1 | - | 1 - 2 | 220 | 64 | |
| AK091956.1 | - | 1 - 2 | 872 | 64 | ||
| AK292557.1 | - | 1 - 2 | 367 | 64 | ||
| AL834289.1 | - | 1 - 2 | 190 | 64 | ||
| BC093764.1 | - | 1 - 2 | 172 | 7939609 | 64 | |
| BC093790.1 | - | 1 - 2 | 171 | 7939635 | 64 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001811.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03032818.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032818.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032818.hg.1 | TC03001811.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000473093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000473093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000473093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032815.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032815.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032815.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032815.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032815.hg.1 | TC03001811.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032811.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032811.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032811.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032811.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032811.hg.1 | TC03001811.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032813.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032813.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032813.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032813.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032813.hg.1 | TC03001811.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032820.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032820.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032820.hg.1 | TC03001811.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032814.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032814.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032814.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032814.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032814.hg.1 | TC03001811.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000473093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000473093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000473093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012527.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193729.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193729.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193729.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193729.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193729.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193729.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193729.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193729.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193729.hg.1 | TC0300012527.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300193731.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193731.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193731.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193731.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193731.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193731.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193731.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193731.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193731.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193731.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193731.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193731.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193731.hg.1 | TC0300012527.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300193736.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193736.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193736.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193736.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193736.hg.1 | TC0300012527.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300193739.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193739.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193739.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193739.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193739.hg.1 | TC0300012527.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | ENST00000473093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | ENST00000473093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193732.hg.1 | TC0300012527.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300193737.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193737.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193737.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193737.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193737.hg.1 | TC0300012527.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | ENST00000473093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | ENST00000309993.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | ENST00000434835.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | ENST00000473093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | ENST00000481989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | ENST00000491050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | NM_001145417.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | NM_018133.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193733.hg.1 | TC0300012527.hg.1 | Clariom D TC |

MSL2
Genomic Map