Assay Details
| Gene Symbol: CWC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016403.3 | NP_057487.2 | 1 - 2 | 116 | 58 | |
| GenBank mRNA | AF110775.1 | - | 1 - 2 | 56 | 58 | |
| AF161497.1 | - | 1 - 2 | 56 | 58 | ||
| AJ250393.1 | - | 1 - 2 | 73 | 58 | ||
| AK225956.1 | - | 1 - 2 | 75 | 58 | ||
| AK314900.1 | - | 1 - 2 | 60 | 58 | ||
| BC006975.1 | - | 1 - 2 | 79 | 58 | ||
| BC032629.1 | - | 1 - 2 | 74 | 58 | ||
| BC040946.1 | - | 1 - 2 | 68 | 6067745 | 58 | |
| BP231785.1 | - | 1 - 2 | 116 | 58 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002204.hg
Additional Information:
This assay is in the 5'UTR region of the AF110775.1,AF161497.1,AJ250393.1,AK314900.1,BC006975.1,BC032629.1,BC040946.1,NM_016403.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000616442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000616442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000616442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000616442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000616442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000616442.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026378.hg.1 | ENST00000545018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026378.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026378.hg.1 | TC11002204.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026376.hg.1 | ENST00000545018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026376.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026376.hg.1 | TC11002204.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026380.hg.1 | ENST00000545018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026380.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026380.hg.1 | TC11002204.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100192564.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192564.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192564.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192564.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192564.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192564.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100192564.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192564.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192564.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192564.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192564.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192564.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100192564.hg.1 | TC1100012012.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100192570.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192570.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100192570.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192570.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100192570.hg.1 | TC1100012012.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000616442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000616442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000616442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000616442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000616442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000616442.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012012.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100192560.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192560.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192560.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192560.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192560.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192560.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100192560.hg.1 | ENST00000279839.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192560.hg.1 | ENST00000539856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192560.hg.1 | ENST00000542165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192560.hg.1 | ENST00000542907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192560.hg.1 | ENST00000621358.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100192560.hg.1 | NM_016403.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100192560.hg.1 | TC1100012012.hg.1 | Clariom D TC |

CWC15
Genomic Map