| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024085.hg.1 |
ENST00000156499.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024085.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024085.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024085.hg.1 |
TC19001771.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024086.hg.1 |
ENST00000156499.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024086.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024086.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024086.hg.1 |
TC19001771.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900194581.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194581.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194581.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194581.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194581.hg.1 |
TC1900011237.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900194581.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194581.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194581.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194581.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000156499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
ENST00000391802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_001311182.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011237.hg.1 |
NM_022046.5 |
Refseq |