Assay Details
| Gene Symbol: AKAP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008534.1 | NP_001008534.1 | 1 - 2 | 126 | 94 | |
| NM_001008535.1 | NP_001008535.1 | 1 - 2 | 126 | 94 | ||
| NM_178813.5 | NP_848928.1 | 1 - 2 | 126 | 94 | ||
| GenBank mRNA | AF514780.1 | - | 1 - 2 | 73 | 94 | |
| AF514781.1 | - | 1 - 2 | 73 | 94 | ||
| AF514782.1 | - | 1 - 2 | 73 | 94 | ||
| BC066357.1 | - | 2 - 3 | 263 | 5295545 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000590.hg
Additional Information:
This assay is in the 5'UTR region of the AF514780.1,AF514781.1,AF514782.1,BC066357.1,NM_001008534.1,NM_001008535.1,NM_178813.5 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007775.hg.1 | ENST00000371423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007775.hg.1 | ENST00000371425.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007775.hg.1 | ENST00000371431.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007775.hg.1 | ENST00000394594.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007775.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007775.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007775.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007775.hg.1 | TC0X000590.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007776.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007776.hg.1 | ENST00000371422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007776.hg.1 | ENST00000371423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007776.hg.1 | ENST00000371425.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007776.hg.1 | ENST00000371431.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007776.hg.1 | ENST00000394594.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007776.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007776.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007776.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007776.hg.1 | TC0X000590.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007777.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007777.hg.1 | ENST00000371422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007777.hg.1 | ENST00000371423.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007777.hg.1 | ENST00000371425.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007777.hg.1 | ENST00000371431.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007777.hg.1 | ENST00000394594.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007777.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007777.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007777.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007777.hg.1 | TC0X000590.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | ENST00000491105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008264.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | ENST00000371422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | ENST00000334356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | ENST00000371422.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | ENST00000371425.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | ENST00000371431.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | NM_001008534.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | NM_001008535.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | NM_178813.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00156468.hg.1 | TC0X00008264.hg.1 | Clariom D TC |

AKAP14
Genomic Map