Assay Details
| Gene Symbol: CATSPERB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024764.3 | NP_079040.2 | 18 - 19 | 2094 | 127 | |
| GenBank mRNA | AK024360.1 | - | 4 - 5 | 622 | 127 | |
| AK126034.1 | - | 18 - 19 | 2078 | 127 | ||
| BC126214.1 | - | 18 - 19 | 2048 | 8991930 | 127 | |
| BC126216.1 | - | 18 - 19 | 2048 | 8991932 | 127 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001439.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14016071.hg.1 | ENST00000256343.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016071.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016071.hg.1 | TC14001439.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000553329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000556901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010776.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183068.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183068.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183068.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183068.hg.1 | ENST00000256343.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183068.hg.1 | ENST00000557036.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183068.hg.1 | NM_024764.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183068.hg.1 | TC1400010776.hg.1 | Clariom D TC |

CATSPERB
Genomic Map