Assay Details
| Gene Symbol: PCED1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271168.1 | NP_001258097.1 | 4 - 5 | 686 | 76 | |
| NM_022760.4 | NP_073597.2 | 4 - 5 | 953 | 76 | ||
| XM_005260804.2 | XP_005260861.1 | 4 - 5 | 839 | 76 | ||
| XM_005260805.2 | XP_005260862.1 | 4 - 5 | 733 | 76 | ||
| XM_005260806.3 | XP_005260863.1 | 4 - 5 | 886 | 76 | ||
| GenBank mRNA | AK026029.1 | - | 4 - 5 | 594 | 76 | |
| AK056353.1 | - | 4 - 5 | 937 | 76 | ||
| AK126535.1 | - | 1 - 2 | 754 | 76 | ||
| AK293638.1 | - | 3 - 4 | 835 | 76 | ||
| AY211913.1 | - | 2 - 3 | 279 | 76 | ||
| BC014247.2 | - | 4 - 5 | 587 | 4641068 | 76 | |
| BC037240.1 | - | 4 - 5 | 559 | 5198102 | 76 | |
| BC051816.1 | - | 4 - 5 | 741 | 5220123 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000566.hg
Additional Information:
This assay is in the 5'UTR region of the AK293638.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008215.hg.1 | ENST00000356872.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008215.hg.1 | ENST00000360652.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008215.hg.1 | ENST00000478539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008215.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008215.hg.1 | TC20000566.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008217.hg.1 | ENST00000356872.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008217.hg.1 | ENST00000360652.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008217.hg.1 | ENST00000380531.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008217.hg.1 | ENST00000478539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008217.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008217.hg.1 | TC20000566.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008214.hg.1 | ENST00000356872.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008214.hg.1 | ENST00000360652.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008214.hg.1 | ENST00000478539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008214.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008214.hg.1 | TC20000566.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000474714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | ENST00000487501.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008200.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | ENST00000487501.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | NM_022760.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | TC2000008200.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | ENST00000356872.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | ENST00000360652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | ENST00000439542.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | ENST00000448755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | ENST00000487501.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | NM_001271168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157786.hg.1 | NM_022760.4 | Refseq |

PCED1A
Genomic Map