| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001871.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001871.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001871.hg.1 |
TC0Y000186.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
ENST00000439472.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
ENST00000439472.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
ENST00000439472.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001870.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001870.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001870.hg.1 |
TC0Y000186.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001869.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001869.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001869.hg.1 |
TC0Y000186.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
ENST00000439472.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
ENST00000439472.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
ENST00000439472.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0Y00007167.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00148248.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00148248.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00148248.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00148248.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00148248.hg.1 |
TC0Y00007167.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0Y00148247.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00148247.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00148247.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00148247.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00148247.hg.1 |
TC0Y00007167.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0Y00148246.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00148246.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00148246.hg.1 |
ENST00000455084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00148246.hg.1 |
NR_001542.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00148246.hg.1 |
TC0Y00007167.hg.1 |
Clariom D TC |