Assay Details
| Gene Symbol: TMEM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305050.1 | NP_001291979.1 | 1 - 2 | 438 | 95 | |
| XM_006711051.1 | XP_006711114.1 | 1 - 2 | 436 | 95 | ||
| GenBank mRNA | BI604139.1 | - | 1 - 2 | 206 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | ENST00000234831.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | ENST00000371337.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | ENST00000452421.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | NM_001305043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | NM_001305050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | NM_001305066.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | NM_004872.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | ENST00000234831.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | ENST00000371337.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | ENST00000452421.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | NM_001305043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | NM_001305050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | NM_001305066.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | NM_004872.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206774.hg.1 | TC0100014250.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | ENST00000234831.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | ENST00000371341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | ENST00000440019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_001305043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_001305049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_001305050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_001305051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_001305052.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_001305066.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_004872.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | ENST00000234831.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | ENST00000371341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | ENST00000440019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_001305043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_001305049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_001305050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_001305051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_001305052.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_001305066.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | NM_004872.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206782.hg.1 | TC0100014250.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000234831.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000371341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000371344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000371348.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000420738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000440019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000452421.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | NM_001305043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | NM_001305049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | NM_001305050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | NM_001305051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | NM_001305052.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | NM_004872.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000234831.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000371341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000371344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000371348.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000420738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000440019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | ENST00000452421.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | NM_001305043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | NM_001305049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | NM_001305050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | NM_001305051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | NM_001305052.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | NM_004872.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206763.hg.1 | TC0100014250.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100206778.hg.1 | ENST00000234831.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206778.hg.1 | ENST00000452421.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206778.hg.1 | NM_001305043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206778.hg.1 | NM_001305050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206778.hg.1 | NM_001305066.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206778.hg.1 | NM_004872.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206778.hg.1 | ENST00000234831.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206778.hg.1 | ENST00000452421.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206778.hg.1 | NM_001305043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206778.hg.1 | NM_001305050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206778.hg.1 | NM_001305066.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206778.hg.1 | NM_004872.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206778.hg.1 | TC0100014250.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100206779.hg.1 | ENST00000234831.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206779.hg.1 | ENST00000452421.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206779.hg.1 | NM_001305043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206779.hg.1 | NM_001305050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206779.hg.1 | NM_001305066.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206779.hg.1 | NM_004872.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206779.hg.1 | ENST00000234831.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206779.hg.1 | ENST00000452421.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206779.hg.1 | NM_001305043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206779.hg.1 | NM_001305050.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206779.hg.1 | NM_001305066.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206779.hg.1 | NM_004872.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206779.hg.1 | TC0100014250.hg.1 | Clariom D TC |

TMEM59
Genomic Map