Assay Details
| Gene Symbol: HMGCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166107.1 | NP_001159579.1 | 1 - 2 | 194 | 83 | |
| NM_005518.3 | NP_005509.1 | 1 - 2 | 194 | 83 | ||
| XM_011541313.1 | XP_011539615.1 | 1 - 2 | 387 | 83 | ||
| GenBank mRNA | AK301594.1 | - | 1 - 2 | 164 | 83 | |
| AK302276.1 | - | 1 - 2 | 167 | 83 | ||
| AK303777.1 | - | 1 - 2 | 167 | 83 | ||
| AK313590.1 | - | 1 - 2 | 169 | 83 | ||
| BC044217.1 | - | 1 - 2 | 147 | 5744956 | 83 | |
| BF964973.1 | - | 1 - 2 | 168 | 83 | ||
| BP420631.1 | - | 1 - 2 | 203 | 83 | ||
| CR456850.1 | - | 1 - 2 | 106 | 83 | ||
| X83618.1 | - | 1 - 2 | 157 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003050.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK301594.1,AK302276.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
kidney development
liver development
cholesterol biosynthetic process
brain development
midgut development
response to nutrient
isoprenoid biosynthetic process
response to temperature stimulus
response to metal ion
lung development
cellular response to insulin stimulus
multicellular organismal response to stress
response to testosterone
response to glucagon
response to triglyceride
response to monosaccharide
response to prostaglandin F
response to drug
response to starvation
response to ethanol
ketone body biosynthetic process
response to cAMP
response to growth hormone
adipose tissue development
response to linoleic acid
cellular response to lipopolysaccharide
cellular response to amino acid stimulus
cellular response to glucocorticoid stimulus
cellular response to fatty acid
hydroxymethylglutaryl-CoA synthase activity
kidney development
liver development
cholesterol biosynthetic process
brain development
midgut development
response to nutrient
isoprenoid biosynthetic process
response to temperature stimulus
response to metal ion
lung development
cellular response to insulin stimulus
multicellular organismal response to stress
response to testosterone
response to glucagon
response to triglyceride
response to monosaccharide
response to prostaglandin F
response to drug
response to starvation
response to ethanol
ketone body biosynthetic process
response to cAMP
response to growth hormone
adipose tissue development
response to linoleic acid
cellular response to lipopolysaccharide
cellular response to amino acid stimulus
cellular response to glucocorticoid stimulus
cellular response to fatty acid
hydroxymethylglutaryl-CoA synthase activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000472375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000472375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000472375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000472375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000472375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000476640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000476640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000476640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000476640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047494.hg.1 | ENST00000369406.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047494.hg.1 | ENST00000476640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047494.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047494.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047494.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047494.hg.1 | TC01003050.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047498.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047498.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047498.hg.1 | TC01003050.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047495.hg.1 | ENST00000369406.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047495.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047495.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047495.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047495.hg.1 | TC01003050.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000472375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000472375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000472375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000472375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000472375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000476640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000476640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000476640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000476640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015476.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215549.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215549.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215549.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215549.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215549.hg.1 | TC0100015476.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100215544.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215544.hg.1 | ENST00000476640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215544.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215544.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215544.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215544.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215544.hg.1 | ENST00000476640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215544.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215544.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215544.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215544.hg.1 | TC0100015476.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100215545.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215545.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215545.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215545.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215545.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215545.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215545.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215545.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215545.hg.1 | TC0100015476.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100215546.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215546.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215546.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215546.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215546.hg.1 | ENST00000369406.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215546.hg.1 | ENST00000544913.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215546.hg.1 | NM_001166107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215546.hg.1 | NM_005518.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215546.hg.1 | TC0100015476.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 110104 | - | - |

HMGCS2
Genomic Map