| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047486.hg.1 |
ENST00000369406.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047486.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047486.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047486.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01047486.hg.1 |
TC01003050.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000472375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000476640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015476.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215536.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215536.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215536.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215536.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215536.hg.1 |
ENST00000369406.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215536.hg.1 |
ENST00000544913.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100215536.hg.1 |
NM_001166107.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215536.hg.1 |
NM_005518.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100215536.hg.1 |
TC0100015476.hg.1 |
Clariom D TC |