| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000341396.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000341396.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000341396.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000341396.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000341396.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000479098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000479098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000618878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000618878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
ENST00000618878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02007202.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02007202.hg.1 |
TC02000461.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02007203.hg.1 |
ENST00000548978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02007203.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02007203.hg.1 |
TC02000461.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02007201.hg.1 |
ENST00000548978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02007201.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02007201.hg.1 |
TC02000461.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000341396.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000341396.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000341396.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000341396.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000341396.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000377566.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000460508.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000479098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000479098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000550249.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000618878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000618878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
ENST00000618878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001009812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013108.hg.1 |
NM_001282430.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200156111.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156111.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156111.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200156111.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156111.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156111.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200156111.hg.1 |
TC0200008087.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008087.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200156108.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156108.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156108.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200156108.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156108.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156108.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200156108.hg.1 |
TC0200008087.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200156109.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156109.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156109.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200156109.hg.1 |
ENST00000548978.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156109.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156109.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200156109.hg.1 |
TC0200008087.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200156110.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156110.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200156110.hg.1 |
ENST00000603175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200156110.hg.1 |
NR_024606.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200156110.hg.1 |
TC0200008087.hg.1 |
Clariom D TC |