| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014640.hg.1 |
ENST00000276449.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014640.hg.1 |
ENST00000522753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014640.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014640.hg.1 |
TC08001141.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000521236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000521236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000521236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000521236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000521236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014639.hg.1 |
ENST00000276449.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014639.hg.1 |
ENST00000522753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014639.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014639.hg.1 |
TC08001141.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800169007.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169007.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169007.hg.1 |
ENST00000521236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169007.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169007.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800169007.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169007.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169007.hg.1 |
ENST00000521236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169007.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169007.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800169007.hg.1 |
TC0800010153.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800169006.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169006.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169006.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169006.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800169006.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169006.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169006.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800169006.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800169006.hg.1 |
TC0800010153.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000276449.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000520114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000521236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000521236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000521236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000521236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000521236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
ENST00000522050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010153.hg.1 |
NM_000349.2 |
Refseq |